Learn More
TARGETMOL CHEMICALS INC GEFITINIB-BASED PROTAC 3 10MG
SDP
Catalog No. 502579763
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Gefitinib-based PROTAC 3 conjugating an EGFR binding element to a VHL ligand via a linker induces EGFR degradation with DC50s of 11.7 nM and 22.3 nM in HCC827(exon 19 del) and H3255 (L858R mutantion) cells respectively[1]. purity: 98%
Catalog No. 50-257-9763 Supplier TARGETMOL CHEMICALS INC Supplier No. T543710MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Gefitinib-based PROTAC 3 conjugating an EGFR binding element to a VHL ligand via a linker induces EGFR degradation with DC50s of 11.7 nM and 22.3 nM in HCC827(exon 19 del) and H3255 (L858R mutantion) cells respectively[1]. purity: 98%