Learn More
Enzo Life Sciences Gefitinib-based PROTAC 3 (1mg). CAS: 2230821-27-7
SDP
Catalog No. 502013723
Encompass_Preferred
Gefitinib-based PROTAC 3 is targeting on EGFR, consisting of Gefitinib as the warhead linked to VHL-recruiting ligand. Purity: ≥99%. Solubility: Soluble in DMSO (75 mg/mL). Insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-3723 Supplier Enzo Life Sciences Supplier No. ENZCHM2610001
Gefitinib-based PROTAC 3 is targeting on EGFR, consisting of Gefitinib as the warhead linked to VHL-recruiting ligand. Purity: ≥99%. Solubility: Soluble in DMSO (75 mg/mL). Insoluble in water. Long Term Storage: -20°C.