Learn More
Cayman Chemical GefItInIb-based PROTAC 3 25mg
SDP
Catalog No. 502937554
Encompass_Preferred
A PROTAC comprised of gefitinib linked to VHL ligand 1; induces degradation of EGFR containing the activating mutation L858R or an exon 19 deletion (DC50s = 22.3 and 11.7 nM, respectively) but does not induce degradation of wild-type EGFR at ≤10 μM in cell-based assays
Catalog No. 50-293-7554 Supplier Cayman Chemical Supplier No. 2859725 MG
A PROTAC comprised of gefitinib linked to VHL ligand 1; induces degradation of EGFR containing the activating mutation L858R or an exon 19 deletion (DC50s = 22.3 and 11.7 nM, respectively) but does not induce degradation of wild-type EGFR at ≤10 μM in cell-based assays