Learn More
Medchemexpress LLC HY-123921 5mg Medchemexpress, Gefitinib-based PROTAC 3 CAS:2230821-27-7 Purity:>98%
SDP
Catalog No. 501932585
Encompass_Preferred
Medchemexpress, HY-123921 5mg Gefitinib-based PROTAC 3 CAS:2230821-27-7 Gefitinib-based PROTAC 3, conjugating an EGFR binding element to a VHL ligand via a linker, induces EGFR degradation with DC 50 s of 11.7 nM and 22.3 nM in HCC827(exon 19 del) and H3255 (L858R mutantion) cells, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2585 Supplier Medchemexpress LLC Supplier No. HY1239215MG
Medchemexpress, HY-123921 5mg Gefitinib-based PROTAC 3 CAS:2230821-27-7 Gefitinib-based PROTAC 3, conjugating an EGFR binding element to a VHL ligand via a linker, induces EGFR degradation with DC 50 s of 11.7 nM and 22.3 nM in HCC827(exon 19 del) and H3255 (L858R mutantion) cells, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.