Learn More
Medchemexpress LLC Gefitinib-d6 | 1228664-49-0 | 98.0% | C22H18D6ClFN4O3 | 1 MG
SDP
Catalog No. 5000358882
Encompass_Preferred
Gefitinib-d6 is the deuterium labeled Gefitinib. Gefitinib (ZD1839) is a potent, selective, and orally active EGFR tyrosine kinase inhibitor with an IC50 of 33 nM. It selectively inhibits EGF-stimulated tumor cell growth (IC50 of 54 nM) and blocks EGF-stimulated EGFR autophosphorylation in tumor cells. Gefitinib also induces autophagy and has antitumour activity. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-58882 Supplier Medchemexpress LLC Supplier No. HY50895S11MG
Gefitinib-d6 is the deuterium labeled Gefitinib. Gefitinib (ZD1839) is a potent, selective, and orally active EGFR tyrosine kinase inhibitor with an IC50 of 33 nM. It selectively inhibits EGF-stimulated tumor cell growth (IC50 of 54 nM) and blocks EGF-stimulated EGFR autophosphorylation in tumor cells. Gefitinib also induces autophagy and has antitumour activity. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS