Learn More
Medchemexpress LLC Gefitinib-d8 | 857091-32-8 | 98.4% | 454.95 | 1 MG
SDP
Catalog No. 5000365235
Encompass_Preferred
Gefitinib-d8 (ZD1839-d8) is a deuterium-labeled Gefitinib. Gefitinib acts as an EGFR tyrosine kinase inhibitor, with an IC50 of 2-37 nM in NR6wtEGFR cells. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules as tracers for quantitation during drug development. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-65235 Supplier Medchemexpress LLC Supplier No. 1MGHY50895S
Gefitinib-d8 (ZD1839-d8) is a deuterium-labeled Gefitinib. Gefitinib acts as an EGFR tyrosine kinase inhibitor, with an IC50 of 2-37 nM in NR6wtEGFR cells. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules as tracers for quantitation during drug development. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs