Learn More
Abcam Gefitinib, EGFR tyrosine kinase inhibitor, 50MG
Catalog No. 502706363
Encompass_Preferred
MW 446.9 Da, Purity >98%. Orally active, potent, selective EGFR (Epidermal growth factor receptor) tyrosine kinase inhibitor (IC₅₀ = 33 nM). Inhibits EGF-stimulated EGFR autophosphorylation and inhibits tumor growth in vivo. Enhances efficacy of cytotoxic agents. Inhibits growth factor production and angiogenesis. Clinically useful anticancer agent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6363 Supplier Abcam Supplier No. AB14205250MG
MW 446.9 Da, Purity >98%. Orally active, potent, selective EGFR (Epidermal growth factor receptor) tyrosine kinase inhibitor (IC₅₀ = 33 nM). Inhibits EGF-stimulated EGFR autophosphorylation and inhibits tumor growth in vivo. Enhances efficacy of cytotoxic agents. Inhibits growth factor production and angiogenesis. Clinically useful anticancer agent.
The product is subject to the following: Abcam Restricted Use Statement