Learn More
Medchemexpress LLC Gefitinib hydrochloride | 184475-55-6 | 99.8% | 500 MG
SDP
Catalog No. 5000357704
Encompass_Preferred
Gefitinib hydrochloride (ZD1839 hydrochloride) is a potent, selective, and orally active inhibitor of EGFR tyrosine kinase, with an IC50 of 33 nM. It specifically inhibits EGF-stimulated tumor cell growth with an IC50 of 54 nM and blocks EGF-stimulated EGFR autophosphorylation in tumor cells. Additionally, Gefitinib hydrochloride has been shown to induce autophagy and possesses antitumour activity.
  • Potent, selective, and orally active EGFR tyrosine kinase inhibitor.
  • Inhibits EGF-stimulated tumor cell growth.
  • Blocks EGF-stimulated EGFR autophosphorylation in tumor cells.
  • Induces autophagy.
  • Demonstrates antitumour activity.
Catalog No. 50-003-57704 Supplier Medchemexpress LLC Supplier No. HY50895A500MG
Gefitinib hydrochloride (ZD1839 hydrochloride) is a potent, selective, and orally active inhibitor of EGFR tyrosine kinase, with an IC50 of 33 nM. It specifically inhibits EGF-stimulated tumor cell growth with an IC50 of 54 nM and blocks EGF-stimulated EGFR autophosphorylation in tumor cells. Additionally, Gefitinib hydrochloride has been shown to induce autophagy and possesses antitumour activity.
  • Potent, selective, and orally active EGFR tyrosine kinase inhibitor.
  • Inhibits EGF-stimulated tumor cell growth.
  • Blocks EGF-stimulated EGFR autophosphorylation in tumor cells.
  • Induces autophagy.
  • Demonstrates antitumour activity.