Learn More
Medchemexpress LLC Gefitinib hydrochloride | 184475-55-6 | 99.9% | C22H25Cl2FN4O3 | 100 MG
SDP
Catalog No. 5000358420
Encompass_Preferred
Gefitinib hydrochloride (ZD1839 hydrochloride) is a potent, selective, and orally active EGFR tyrosine kinase inhibitor with an IC50 of 33 nM. It selectively inhibits EGF-stimulated tumor cell growth (IC50 of 54 nM) and blocks EGF-stimulated EGFR autophosphorylation in tumor cells. Gefitinib hydrochloride also induces autophagy and has antitumour activity.
  • Potent, selective, and orally active EGFR tyrosine kinase inhibitor
  • Selectively inhibits EGF-stimulated tumor cell growth
  • Blocks EGF-stimulated EGFR autophosphorylation
  • Induces autophagy
  • Has antitumour activity
Catalog No. 50-003-58420 Supplier Medchemexpress LLC Supplier No. HY50895A100MG
Gefitinib hydrochloride (ZD1839 hydrochloride) is a potent, selective, and orally active EGFR tyrosine kinase inhibitor with an IC50 of 33 nM. It selectively inhibits EGF-stimulated tumor cell growth (IC50 of 54 nM) and blocks EGF-stimulated EGFR autophosphorylation in tumor cells. Gefitinib hydrochloride also induces autophagy and has antitumour activity.
  • Potent, selective, and orally active EGFR tyrosine kinase inhibitor
  • Selectively inhibits EGF-stimulated tumor cell growth
  • Blocks EGF-stimulated EGFR autophosphorylation
  • Induces autophagy
  • Has antitumour activity