Learn More
Medchemexpress LLC Gefitinib hydrochloride | 184475-55-6 | C22H25Cl2FN4O3 | 1 ML
SDP
Catalog No. 5000358388
Encompass_Preferred
Gefitinib hydrochloride is a potent, selective, and orally active EGFR tyrosine kinase inhibitor with an IC50 of 33 nM. It selectively inhibits EGF-stimulated tumor cell growth and blocks EGF-stimulated EGFR autophosphorylation in tumor cells.
  • Induces autophagy
  • Has antitumour activity
Catalog No. 50-003-58388 Supplier Medchemexpress LLC Supplier No. HY50895A10MM/1ML
Gefitinib hydrochloride is a potent, selective, and orally active EGFR tyrosine kinase inhibitor with an IC50 of 33 nM. It selectively inhibits EGF-stimulated tumor cell growth and blocks EGF-stimulated EGFR autophosphorylation in tumor cells.
  • Induces autophagy
  • Has antitumour activity