Learn More
Medchemexpress LLC Gefitinib hydrochloride | 184475-55-6 | MFCD00952718 | 99.8% | C22H25Cl2FN4O3 | 1 G
SDP
Catalog No. 5000357751
Encompass_Preferred
Gefitinib hydrochloride is a potent, selective, and orally active EGFR tyrosine kinase inhibitor, intended for research use only. It selectively inhibits EGF-stimulated tumor cell growth and blocks EGFR autophosphorylation in tumor cells. It also induces autophagy and exhibits antitumour activity.
  • Potent and selective EGFR tyrosine kinase inhibitor.
  • IC50 of 33 nM against EGFR.
  • Selectively inhibits EGF-stimulated tumor cell growth with an IC50 of 54 nM.
  • Blocks EGF-stimulated EGFR autophosphorylation in tumor cells.
  • Induces autophagy.
  • Exhibits antitumour activity.
Catalog No. 50-003-57751 Supplier Medchemexpress LLC Supplier No. HY50895A1G
Gefitinib hydrochloride is a potent, selective, and orally active EGFR tyrosine kinase inhibitor, intended for research use only. It selectively inhibits EGF-stimulated tumor cell growth and blocks EGFR autophosphorylation in tumor cells. It also induces autophagy and exhibits antitumour activity.
  • Potent and selective EGFR tyrosine kinase inhibitor.
  • IC50 of 33 nM against EGFR.
  • Selectively inhibits EGF-stimulated tumor cell growth with an IC50 of 54 nM.
  • Blocks EGF-stimulated EGFR autophosphorylation in tumor cells.
  • Induces autophagy.
  • Exhibits antitumour activity.