Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gefitinib impurity 2 | 246512-44-7 | MFCD26395622 | 99.6% | 309.37 g/mol | C15H23N3O4 | 10 MG
SDP

Catalog No. 5000213842
Encompass_Preferred

Gefitinib impurity 2 is a characterized reference impurity of the EGFR inhibitor gefitinib intended for research and analytical use. It is supplied as a solid and as DMSO solutions, and is provided with measured purity and recommended storage conditions to support stability during handling and analysis.

  • High purity: 99.56%.
  • Molecular formula: C15H23N3O4.
  • Molecular weight: 309.37 g/mol.
  • Appearance: off-white to pink solid.
  • Available formats: 10 mg solid, 5 mg solid, 10 mM 1 mL DMSO solution.
  • Storage recommendations: powder at -20 °C for long term; in solvent at -80 °C for extended stability.

Catalog No. 50-002-13842 Supplier Medchemexpress LLC Supplier No. HY10066310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gefitinib impurity 2 is a characterized reference impurity of the EGFR inhibitor gefitinib intended for research and analytical use. It is supplied as a solid and as DMSO solutions, and is provided with measured purity and recommended storage conditions to support stability during handling and analysis.

  • High purity: 99.56%.
  • Molecular formula: C15H23N3O4.
  • Molecular weight: 309.37 g/mol.
  • Appearance: off-white to pink solid.
  • Available formats: 10 mg solid, 5 mg solid, 10 mM 1 mL DMSO solution.
  • Storage recommendations: powder at -20 °C for long term; in solvent at -80 °C for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.