Learn More
Selleck Chemical LLC Gefitinib (ZD1839) 100mg 184475-35-2 Iressa
SDP
Supplier:  Selleck Chemical LLC S1025100MG
Encompass_Preferred
Gefitinib (ZD-1839, Iressa) is an EGFR inhibitor for Tyr1173, Tyr992, Tyr1173 and Tyr992 in the NR6wtEGFR and NR6W cells with IC50 of 37 nM, 37nM, 26 nM and 57 nM, respectively. Gefitinib promotes autophagy and apoptosis of lung cancer cells via blockade of the PI3K/AKT/mTOR pathway. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-778-5