Learn More
MSC Geier & Bluhm Bull's Eye Circular Level, 1-1/4 in Base Diameter, Brass
Catalog No. 5000528050
Encompass
This circular level features a brass construction with a 1-1/4 inch base diameter and 0.44 inch overall height. Designed for precise leveling, it offers a sensitivity of 45 minutes and operates effectively within a wide temperature range.
  • Brass construction for durability
  • 1-1/4 inch base diameter
  • 0.44 inch overall height
  • 0.12 inch mounting hole diameter
  • 0.276 inch vial diameter
  • 45 minute sensitivity
  • Operates within a temperature range of -40 to 150 °F
  • Made in the United States
  • Backed by a hassle-free guarantee for returns within 30 days
Catalog No. 50-005-28050 Supplier MSC Supplier No. 76482066
This circular level features a brass construction with a 1-1/4 inch base diameter and 0.44 inch overall height. Designed for precise leveling, it offers a sensitivity of 45 minutes and operates effectively within a wide temperature range.
  • Brass construction for durability
  • 1-1/4 inch base diameter
  • 0.44 inch overall height
  • 0.12 inch mounting hole diameter
  • 0.276 inch vial diameter
  • 45 minute sensitivity
  • Operates within a temperature range of -40 to 150 °F
  • Made in the United States
  • Backed by a hassle-free guarantee for returns within 30 days