Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Geiger Counter, Digital Readout
SDPSCI_ED

Catalog No. s441001
Change view
Click to view available options
Display Type:
LCD
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Type
S441001 LCD
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S441001 Supplier Science First Supplier No. 10810

Includes

AC adapter for 110VAC line, laboratory manual with a wide range of experiments, demonstrations and field investigations

Requires

Four AA batteries (S43921)

Compact, sturdy, digital instrument for field or lab radiation measurement.

  • Halogen-quenched, stainless-steel Geiger Müller tube Sturdy and lightweight construction Operates on AC line voltage or with batteries
  • Sensitive to low-level detection of alpha, beta and gamma radiation
  • Digital readout can be converted to mR/hr. using the scale factor on the unit
  • Can take a count over 7.5 seconds or at a 60-second interval

Order Info

Dimensions (approx.): 7.1 x 3.9 x 2.4 in. (18 x 10 x 6cm).

Specifications

Sensitivity 18 cps/mR/Hour (Co60), >0.01 MeV Gamma, >0.2MeV beta, >4 MeV alpha
Display Type LCD
Battery Type 2 AA
Product Type Digital Geiger Counter
Measurement Range 0 to 8 mR/Hour
Detector Type Halogen-quenched Ne stainless steel geiger mueller tube
Active Area 1.5 to 2 mg/cm2 Mica
For Use With (Application) For field and laboratory use
Voltage 500 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.