Learn More
Geiger Detector with Switch
SDPSCI_ED
Catalog No. s441002
Change view
Click to view available options
Geiger Counter, Plug-in
Catalog No.
S441002 Geiger Counter, Plug-in
1 options
This item is not returnable. View return policy
Catalog No. S441002 Supplier Science First Supplier No. 10609
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
A self-contained and AC powered detector and speaker which produces a sharp click for each event when exposed to background radiation or sources of alpha, beta, or gamma radiation. Useful for qualitative comparisons, this unit can show the effects of distance, shielding, and activity of a radiation source.
  • Plugs into a standard 115VAC outlet
  • Specially designed to give a sharp audible click for counting
  • Unit has a Geiger tube window on the right side and a speaker on the front
  • Meter detects alpha, beta, and gamma radiation and is sensitive to background radiation
  • Simple radiation detector will support a range of radiation demonstrations and experiments
Specifications
Product Type Geiger Counter, Plug-in
For Use With (Application) Radioactive demonstrations and experiment
Includes Switch