Learn More
Medchemexpress LLC HY-N2411 5mg Medchemexpress, Geissoschizine methyl ether CAS:60314-89-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN24115MG
Encompass_Preferred
Medchemexpress, HY-N2411 5mg Geissoschizine methyl ether CAS:60314-89-8 Geissoschizine methyl ether, a major indole alkaloid found in Uncaria hook, is a major active component of Yokukansan with psychotropic effects. Geissoschizine methyl ether is potent 5-HT1A receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2894