Learn More
Bio Rad Laboratories GEL BIO-GEL P-100
Catalog No. 509010009
Encompass
Bio-Gel P-100 Polyacrylamide Beads, Medium polyacrylamide beads for size exclusion chromatography, 90-180um wet bead size, 5,000-100,000 MW fractionation range, 100g
Catalog No. 50-901-0009 Supplier Bio Rad Laboratories Supplier No. 1504170
Bio-Gel P-100 Polyacrylamide Beads, Medium polyacrylamide beads for size exclusion chromatography, 90-180um wet bead size, 5,000-100,000 MW fractionation range, 100g