Learn More
Bio Rad Laboratories GEL BIO-GEL P-2
Catalog No. 509012913
Encompass
Gel; Bio-Rad; Bio-Gel; P-2; Polyacrylamide media; Hydrophilic; Nonionic; Autoclavable at pH 5.5-6.5; Operate over a pH range of 2-10 at room temperature; Extra fine; 100g
Catalog No. 50-901-2913 Supplier Bio Rad Laboratories Supplier No. 1504118
Gel; Bio-Rad; Bio-Gel; P-2; Polyacrylamide media; Hydrophilic; Nonionic; Autoclavable at pH 5.5-6.5; Operate over a pH range of 2-10 at room temperature; Extra fine; 100g