Learn More
Bio Rad Laboratories GEL BIO-GEL P-30
Supplier:  Bio Rad Laboratories 1504154
Encompass
Gel; Bio-Rad; Bio-Gel; P-30; Polyacrylamide media; Hydrophilic; Nonionic; Autoclavable at pH 5.5-6.5; Operate over a pH range of 2-10 at room temperature; Fine; 100g
Catalog No. 50-901-2918