Learn More
Hoefer, Inc GEL BOX W/COOL BASE 20X20CM
SDP
Catalog No. NC3902383
Encompass_Preferred
SUBMARINE HORIZONTEL GEL BOX WITH COOLING BASE FOR RUNNING MAXI NUCLEIC ACID GELS 20X20CM.INCLUDES TWO 1MM THICK 16WELL COMBS
Catalog No. NC3902383 Supplier Hoefer, Inc Supplier No. SUB20C
SUBMARINE HORIZONTEL GEL BOX WITH COOLING BASE FOR RUNNING MAXI NUCLEIC ACID GELS 20X20CM.INCLUDES TWO 1MM THICK 16WELL COMBS