Learn More
Ist Engineering Gel Breaker Tubes (Bag of 100)
Catalog No. NC0462125
Encompass
• Patent pending design fragments gel slices into small pieces for easy extraction.• Allows simple transfer using 1 mL pipet tips without clogging.The gel breaker tube is a 0.5 mL tube with four 10μm holes designed to fragment acrylamide and agarose gel slices. The gel slice is placed inside the gel breaker tube which is then stacked into a 2 mL microfuge tube. Centrifugation at 14,000 rpm allows the gel to move through the holes, fragmenting the gel. Water or buffer can then be added to elute out the DNA or RNA. The fragmented gel pieces are small enough that a 1 mL pipet tip will pick up the gel pieces, allowing easy transfer to the 5μm filter tubes.
Catalog No. NC0462125 Supplier Ist Engineering Supplier No. 3388100
• Patent pending design fragments gel slices into small pieces for easy extraction.• Allows simple transfer using 1 mL pipet tips without clogging.The gel breaker tube is a 0.5 mL tube with four 10μm holes designed to fragment acrylamide and agarose gel slices. The gel slice is placed inside the gel breaker tube which is then stacked into a 2 mL microfuge tube. Centrifugation at 14,000 rpm allows the gel to move through the holes, fragmenting the gel. Water or buffer can then be added to elute out the DNA or RNA. The fragmented gel pieces are small enough that a 1 mL pipet tip will pick up the gel pieces, allowing easy transfer to the 5μm filter tubes.