Learn More
Alkali Scientific Gel Breaker Tubes – Spin Columns with 600µm Pores, 200/pk
SDP
Catalog No. 5000662075
Encompass_Preferred
Gel Crusher™ Spin Columns are high performance gel breaker tubes designed to simplify and accelerate gel fragmentation workflows in molecular biology laboratories. Each column contains ten large 600µm filtration pores that efficiently break down agarose or acrylamide gel slices during centrifugation. The column fits securely into a standard 2mL microcentrifuge receiver tube, allowing rapid processing of gel fragments. When centrifuged at high speed, the gel is forced through the pores, producing uniform fragments that are easily pipetted using wide bore tips. The resulting fragments absorb buffer more quickly, supporting improved nucleic acid recovery and more efficient downstream workflows such as cloning, PCR, restriction digestion, and nucleic acid purification.
Catalog No. 50-006-62075 Supplier Alkali Scientific Supplier No. GCSC2ML
Gel Crusher™ Spin Columns are high performance gel breaker tubes designed to simplify and accelerate gel fragmentation workflows in molecular biology laboratories. Each column contains ten large 600µm filtration pores that efficiently break down agarose or acrylamide gel slices during centrifugation. The column fits securely into a standard 2mL microcentrifuge receiver tube, allowing rapid processing of gel fragments. When centrifuged at high speed, the gel is forced through the pores, producing uniform fragments that are easily pipetted using wide bore tips. The resulting fragments absorb buffer more quickly, supporting improved nucleic acid recovery and more efficient downstream workflows such as cloning, PCR, restriction digestion, and nucleic acid purification.