Learn More
Gel Company GEL CUTTER BLADES 2 MM X 9 MM
Catalog No. NC0623333
Encompass
Disposable gridcutters simultaneously cut and excise 34 gel slices from a gel. Each slice is approx 2 mm x 9 mm. Ship as 1 pack of 10. Blades are attached to a mount (sold separately, see cat # MEF67-9).
Catalog No. NC0623333 Supplier Gel Company Supplier No. MEE2934
Disposable gridcutters simultaneously cut and excise 34 gel slices from a gel. Each slice is approx 2 mm x 9 mm. Ship as 1 pack of 10. Blades are attached to a mount (sold separately, see cat # MEF67-9).