Learn More
Midwest Scientific Axygen 11 mm x 4 mm Gel Cutting Pipet Tips, Clear, Nonsterile, Rack Pack, 250 Tips/Rack, 10 Packs/Case
SDP
Catalog No. NC1069970
Encompass
Axygen Gel Cutting Tips are designed to remove bands from agarose gel. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. Designed for removing bands from agarose gel One handed operation Push button gel release Ejector button tip release Avoid cross contamination Tips fit standard 1000L pipettors These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. Nonpyrogenic Nonsterile
Catalog No. NC1069970 Supplier Midwest Scientific Supplier No. TGL1140
Axygen Gel Cutting Tips are designed to remove bands from agarose gel. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. Designed for removing bands from agarose gel One handed operation Push button gel release Ejector button tip release Avoid cross contamination Tips fit standard 1000L pipettors These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. Nonpyrogenic Nonsterile