Learn More
Med Vet International Diagel Diarrhea Control Gel For Large Dogs, 61 Pounds Or More, 5mL, Each
Catalog No. 502828657
Encompass_Preferred
An orally administered product to aid in controlling bacterial diarrhea by killing bacteria such as E. coli and Salmonella in the GI tract. Active ingredient, Axaphen, is a patented compound that promotes gut health and eliminates bacteria on contact. 100% product guarantee.
Catalog No. 50-282-8657 Supplier Med Vet International Supplier No. PHDIAGELDOGL
An orally administered product to aid in controlling bacterial diarrhea by killing bacteria such as E. coli and Salmonella in the GI tract. Active ingredient, Axaphen, is a patented compound that promotes gut health and eliminates bacteria on contact. 100% product guarantee.