Learn More
Electron Microscopy Sciences Gel Drying Frames 14 x 14cm Gel Frame
SDP
Catalog No. 5034528
Encompass_Preferred
Gel Drying Frames allow researchers to quickly and easily dry gels between two sheets of cellophane without the use of expensive heat/vacuum gel dryers. 14x14cm frames are ideal for mini gels and smaller formats. Kit: 2 open frames, 2 solid back plates, 12 side clips, 50 sheets pre-cut cellophane.
Catalog No. 50-345-28 Supplier Electron Microscopy Sciences Supplier No. 6851014
Gel Drying Frames allow researchers to quickly and easily dry gels between two sheets of cellophane without the use of expensive heat/vacuum gel dryers. 14x14cm frames are ideal for mini gels and smaller formats. Kit: 2 open frames, 2 solid back plates, 12 side clips, 50 sheets pre-cut cellophane.