Learn More
Sigma Aldrich Fine Chemicals Biosciences Gel Filtration Cal Kit Hig
Catalog No. 502908055
Encompass
The High Molecular Weight (HMW) Kit contains five proteins with Mr in the range 43 000 to 669 000 and Blue Dextran 2000 and provides simple reliable calibration of gel filtration columns.The kits are suitable for calibration of high-reSOLUTIONon gel filtration media and columns such as Superdex Superose and Sephacryl.
Catalog No. 50-290-8055 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28403842
The High Molecular Weight (HMW) Kit contains five proteins with Mr in the range 43 000 to 669 000 and Blue Dextran 2000 and provides simple reliable calibration of gel filtration columns.The kits are suitable for calibration of high-reSOLUTIONon gel filtration media and columns such as Superdex Superose and Sephacryl.