Learn More
Sigma Aldrich Fine Chemicals Biosciences Gel filtration Markers Kit
Catalog No. 502904402
Encompass
Gel filtration markers kit contains five vials of proteins and 1 vial of blue dextran for calibration.
Catalog No. 50-290-4402 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MWGF2001KT
Gel filtration markers kit contains five vials of proteins and 1 vial of blue dextran for calibration.