Learn More
Biotage GEL FILTRATION PHYTIP COLUMNS
SDP
Catalog No. NC2126784
Encompass
Gel filtration PhyTip® columns (1000 µL volume) containing 600 µL of 5K size exclusion resin per column
This item is not returnable. View return policy
Catalog No. NC2126784 Supplier Biotage Supplier No. PDR916006
Gel filtration PhyTip® columns (1000 µL volume) containing 600 µL of 5K size exclusion resin per column