Learn More
Bio Rad Laboratories GEL FILTRATION STD W/6 VIALS
Catalog No. NC9255856
Encompass
Bio Rad No.: 1511901; Product: Gel Filtration Standard, 6 vials, Contains 5mg each of Thyroglobulin, Bovine gamma globulin, and Chicken ovalbumin, 2.5mg Equine myoglobin, and 0.5mg Vitamin B-12
Catalog No. NC9255856 Supplier Bio Rad Laboratories Supplier No. 1511901
Bio Rad No.: 1511901; Product: Gel Filtration Standard, 6 vials, Contains 5mg each of Thyroglobulin, Bovine gamma globulin, and Chicken ovalbumin, 2.5mg Equine myoglobin, and 0.5mg Vitamin B-12