Learn More
Biotium GelGreen Nucleic Acid Gel Stain 10,000x in DMSO
Catalog No. NC9953067
Encompass_Preferred
GelGreen is a sensitive, stable and environmentally safer alternative to the highly toxic ethidium bromide (EB) for staining dsDNA, ssDNA or RNA in agarose or polyacrylamide gels. The dye is noncytotoxic, nonmutagenic and nonhazardous at concentrations used in gel staining. It can be conveniently disposed down the drain or in regular trash at working concentrations, reducing disposal costs. It is much more sensitive than EB or SYBR Safe and extremely stable for long-term room temperature storage. Simple to use in precast or post-electrophoresis gel staining. Compatible with a UV transilluminator, gel reader or scanner with visible light, blue LED, or 488 nm laser-based excitation; replaces SYBR with no optical setting change. Compatible with downstream gel purification, restriction digest, sequencing and cloning. See also GelRed Nucleic Acid Gel Stain, our safer replacement for EB.
Catalog No. NC9953067 Supplier Biotium Supplier No. 41004
GelGreen is a sensitive, stable and environmentally safer alternative to the highly toxic ethidium bromide (EB) for staining dsDNA, ssDNA or RNA in agarose or polyacrylamide gels. The dye is noncytotoxic, nonmutagenic and nonhazardous at concentrations used in gel staining. It can be conveniently disposed down the drain or in regular trash at working concentrations, reducing disposal costs. It is much more sensitive than EB or SYBR Safe and extremely stable for long-term room temperature storage. Simple to use in precast or post-electrophoresis gel staining. Compatible with a UV transilluminator, gel reader or scanner with visible light, blue LED, or 488 nm laser-based excitation; replaces SYBR with no optical setting change. Compatible with downstream gel purification, restriction digest, sequencing and cloning. See also GelRed Nucleic Acid Gel Stain, our safer replacement for EB.