Learn More
Electron Microscopy Sciences EMS SmartDoc™ Gel Imaging System with Blue Light Illumination, 230V
SDP
Catalog No. 501927932
Encompass_Preferred
The SmartDoc system uses a smart phone or tablet to quickly obtain quality images of gels for your records or anaylsis. With an adjustable focal distance for optimum focus, the SmartDoc enclosure is simply place over your gel and transilluminator. Lay your smart phone or tablet onto the non-slip, rubber pad, and align the camera lens over the filter. Raising and lowering the platform allows optimization of focus and image size. Filters are available for compatibility with multiple light sources and dyes. The Blue Light model uses high intensity blue LEDs to excite green dyes and this eliminates the need for hazardous and DNA-damaging UV light.Images can be previewed on the device's display before capturing the picture. Images are easily shared, printed or transferred via text message, email or cloud sharing apps. Gel analysis software can be used to analyze the image. Includes high intensity blue LEDs to excite green dyes that eliminates UV light.
Catalog No. 50-192-7932 Supplier Electron Microscopy Sciences Supplier No. 8705703
The SmartDoc system uses a smart phone or tablet to quickly obtain quality images of gels for your records or anaylsis. With an adjustable focal distance for optimum focus, the SmartDoc enclosure is simply place over your gel and transilluminator. Lay your smart phone or tablet onto the non-slip, rubber pad, and align the camera lens over the filter. Raising and lowering the platform allows optimization of focus and image size. Filters are available for compatibility with multiple light sources and dyes. The Blue Light model uses high intensity blue LEDs to excite green dyes and this eliminates the need for hazardous and DNA-damaging UV light.Images can be previewed on the device's display before capturing the picture. Images are easily shared, printed or transferred via text message, email or cloud sharing apps. Gel analysis software can be used to analyze the image. Includes high intensity blue LEDs to excite green dyes that eliminates UV light.