Learn More
New England Biolabs, Inc. Gel Loading Dye Blue (6X) – 4 ml
SDP
Catalog No. 50994905
Encompass
Gel Loading Dye, Blue (6X) is a Bromophenol Blue-based loading dye offering convenient gel loading and sharp bands. You may also be interested in our Gel Loading Dye, Purple (6X) with and without SDS, for sharp bands and no UV shadow. Gel Loading Dye, Blue (6X) is a pre-mixed loading buffer with one tracking dye for agarose and non-denaturing polyacrylamide gel electrophoresis. This solution contains SDS, which often results in sharper bands, as some restriction enzymes are known to remain bound to DNA following cleavage. EDTA is also included to chelate magnesium (up to 10 mM) in enzymatic reactions, thereby stopping the reaction. Bromophenol Blue is the standard tracking dye for electrophoresis. It migrates at approximately 300 bp on a standard 1% TBE agarose gel. This product is packaged as 4x1 ml vials.
Catalog No. 50-994-905 Supplier New England Biolabs, Inc. Supplier No. B7021S
Gel Loading Dye, Blue (6X) is a Bromophenol Blue-based loading dye offering convenient gel loading and sharp bands. You may also be interested in our Gel Loading Dye, Purple (6X) with and without SDS, for sharp bands and no UV shadow. Gel Loading Dye, Blue (6X) is a pre-mixed loading buffer with one tracking dye for agarose and non-denaturing polyacrylamide gel electrophoresis. This solution contains SDS, which often results in sharper bands, as some restriction enzymes are known to remain bound to DNA following cleavage. EDTA is also included to chelate magnesium (up to 10 mM) in enzymatic reactions, thereby stopping the reaction. Bromophenol Blue is the standard tracking dye for electrophoresis. It migrates at approximately 300 bp on a standard 1% TBE agarose gel. This product is packaged as 4x1 ml vials.