Learn More
Alkali Scientific Gel Loading Flat Tips, 10uL 52mm Length, Outer Diameter: 0.37 mm, 960 tips/Case, Sterile
SDP
Catalog No. 5000663206
Encompass_Preferred
Gel Loading Flat Tips, 10 µL, 52 mm length are designed for precise and controlled sample loading in electrophoresis applications. The flat tip design with a narrow 0.37 mm outer diameter enables accurate delivery of samples into small or closely spaced gel wells while minimizing sample loss and cross contamination. The extended 52 mm length allows convenient access to deep or narrow wells, supporting smooth and controlled dispensing. These tips are suitable for DNA, RNA, and protein gel loading workflows where precision and consistency are essential. Precision molded construction ensures reliable performance, and the non filtered design makes them appropriate for standard electrophoresis procedures in research laboratories.
Catalog No. 50-006-63206 Supplier Alkali Scientific Supplier No. RGLF10S
Gel Loading Flat Tips, 10 µL, 52 mm length are designed for precise and controlled sample loading in electrophoresis applications. The flat tip design with a narrow 0.37 mm outer diameter enables accurate delivery of samples into small or closely spaced gel wells while minimizing sample loss and cross contamination. The extended 52 mm length allows convenient access to deep or narrow wells, supporting smooth and controlled dispensing. These tips are suitable for DNA, RNA, and protein gel loading workflows where precision and consistency are essential. Precision molded construction ensures reliable performance, and the non filtered design makes them appropriate for standard electrophoresis procedures in research laboratories.