Learn More
Alkali Scientific Gel Loading Flat Tips, 200uL 83mm Length, Outer Diameter: 0.17 mm, 960 tips/Case, DNase, RNase & Pyrogen Free
SDP
Catalog No. 5000663329
Encompass_Preferred
Gel Loading Flat Tips, 200 µL, 83 mm Length are designed for precise, large volume sample loading in electrophoresis applications. The ultra fine flat tip with a 0.17 mm outer diameter enables accurate placement of samples into very small or tightly spaced gel wells, helping reduce sample loss and cross contamination. The 83 mm extra long design allows easy access to deep or narrow wells and supports smooth dispensing during DNA, RNA, and protein gel loading procedures. Precision molded construction ensures consistent performance and compatibility with standard micropipettes. Supplied 960 tips per case, these tips are suitable for high throughput electrophoresis workflows in research and molecular biology laboratories.
Catalog No. 50-006-63329 Supplier Alkali Scientific Supplier No. RGLF102
Gel Loading Flat Tips, 200 µL, 83 mm Length are designed for precise, large volume sample loading in electrophoresis applications. The ultra fine flat tip with a 0.17 mm outer diameter enables accurate placement of samples into very small or tightly spaced gel wells, helping reduce sample loss and cross contamination. The 83 mm extra long design allows easy access to deep or narrow wells and supports smooth dispensing during DNA, RNA, and protein gel loading procedures. Precision molded construction ensures consistent performance and compatibility with standard micropipettes. Supplied 960 tips per case, these tips are suitable for high throughput electrophoresis workflows in research and molecular biology laboratories.