Learn More
Alkali Scientific Gel Loading Flat Tips, 200uL 83mm Length, Outer Diameter: 0.37 mm, 960 tips/Case, Sterile
SDP
Catalog No. 5000663217
Encompass_Preferred
Gel Loading Flat Tips, 200 µL, 83 mm Length are designed for accurate, large volume sample loading in electrophoresis workflows. The flat tip with a 0.37 mm outer diameter allows reliable sample delivery into small or moderately spaced gel wells while minimizing sample loss and cross contamination. The extra long 83 mm design provides improved reach into deep or narrow wells and supports controlled dispensing during DNA, RNA, and protein gel loading. Precision molded construction ensures consistent performance and compatibility with standard laboratory micropipettes. Supplied 960 tips per case, these gel loading tips are suitable for routine electrophoresis and molecular biology applications.
Catalog No. 50-006-63217 Supplier Alkali Scientific Supplier No. RGLF237ST
Gel Loading Flat Tips, 200 µL, 83 mm Length are designed for accurate, large volume sample loading in electrophoresis workflows. The flat tip with a 0.37 mm outer diameter allows reliable sample delivery into small or moderately spaced gel wells while minimizing sample loss and cross contamination. The extra long 83 mm design provides improved reach into deep or narrow wells and supports controlled dispensing during DNA, RNA, and protein gel loading. Precision molded construction ensures consistent performance and compatibility with standard laboratory micropipettes. Supplied 960 tips per case, these gel loading tips are suitable for routine electrophoresis and molecular biology applications.