Learn More
Andwin Scientific Gel Loading Tip, Round, Universal Fit, 0.57mm, Volume Capacity: 5 to 200microliter, Length: 83mm, Qty.: 200/Pk.
SDP
Catalog No. NC9826204
Encompass_Preferred
Gel Loading Tip, Round, Universal Fit, 0.57mm, Volume Capacity: 5 to 200microliter, Length: 83mm, Qty.: 200/Pk.
Catalog No. NC9826204 Supplier Andwin Scientific Supplier No. P32051
Gel Loading Tip, Round, Universal Fit, 0.57mm, Volume Capacity: 5 to 200microliter, Length: 83mm, Qty.: 200/Pk.