Learn More
MTC Bio LoadWell™ Gel Loading Pipet Tip, 200ul, 0.7mm diameter tip, Sterile, 10 racks of 96 tips, 960 tips/pk
Catalog No. 502268483
Encompass_Preferred
LoadWell™ Gel Loading Tips have a 0.7mm outer diameter microcapillary orifice, ideal for introducing samples into gel electrophoresis wells. Designed to fit all standard 200µL pipettes (as well as 20µL, 50µL and 100µL pipettes), they are supplied sterile (e-beam), and packaged in autoclavable racks of 96 tips. Produced from virgin, medical grade polypropylene, they are RNase, DNase, endotoxin, and pyrogen free.
This item is not returnable. View return policy
Catalog No. 50-226-8483 Supplier MTC Bio Supplier No. P5720
LoadWell™ Gel Loading Tips have a 0.7mm outer diameter microcapillary orifice, ideal for introducing samples into gel electrophoresis wells. Designed to fit all standard 200µL pipettes (as well as 20µL, 50µL and 100µL pipettes), they are supplied sterile (e-beam), and packaged in autoclavable racks of 96 tips. Produced from virgin, medical grade polypropylene, they are RNase, DNase, endotoxin, and pyrogen free.