Learn More
Alkali Scientific Gel Loading Tips, 200µl, 83mm Length, Round Bore Opening of 0.57mm, 2 Racks of 96 Tips, 5 Inner Packs, 960 Tips/cs
SDP
Supplier:  Alkali Scientific RGL250
Encompass_Preferred
The all for complete visibility of sub-microliter volumes and feature a standard bore. These tips are 100% polypropylene. Not autoclavable. Designed for precise and accurate sample loading into gels. Patented press-to-open racks provide easy, one-handed opening. Universal fit to accommodate most pipettor brands. Multichannel-friendly with a standard 96-tip arrangement. For use in pharmaceutical and food industry, and molecular biology. Manufactured from high-quality, virgin polypropylene to ensure purity and provide excellent optical clarity. Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free.
Catalog No. 50-233-6127