Learn More
Med Vet International Gel Multi-Task Positioner 6"L X 3"W X 3"H, J1282D, Each
Catalog No. 5000581282
Encompass_Preferred
Gel Positioners are designed to aid in positioning patients for either surgery or radiographs. They provide firm but comfortable support. - Made of viscoelastic polymer so it has a soft tissue-like feel - Provides constant support and will not bottom out - Free from latex, silicon, and plasticizer - Will not leak or flow - Radiolucent - Reusable - Easy to clean with standard disinfectants - Compatible with warming blankets - 2 year warranty Flat Bottom Positioner. (Shaped like a loaf of bread) Flat Bottom. 6"L x 3"W x 3"H Note: Gel positioners are not warranted against damage caused by chewing or nail tears.
Catalog No. 50-005-81282 Supplier Med Vet International Supplier No. JOR1282D
Gel Positioners are designed to aid in positioning patients for either surgery or radiographs. They provide firm but comfortable support. - Made of viscoelastic polymer so it has a soft tissue-like feel - Provides constant support and will not bottom out - Free from latex, silicon, and plasticizer - Will not leak or flow - Radiolucent - Reusable - Easy to clean with standard disinfectants - Compatible with warming blankets - 2 year warranty Flat Bottom Positioner. (Shaped like a loaf of bread) Flat Bottom. 6"L x 3"W x 3"H Note: Gel positioners are not warranted against damage caused by chewing or nail tears.