Learn More
Ted Pella Inc GEL PAK STICKY BOXES PK/50
SDP
Catalog No. NC9035618
Encompass
Gel-Pak Clear Hinged Box, GEL Retention X4, 2.54cm x 2.54cm x 0.64cm (1 in. x 1 in. x 1/4 in.), 50/PK, Applications: Semiconductors, thin Film Heads, Microwave Devices, Micromachine Sensors, Optoelectronics, Medical Devices
Catalog No. NC9035618 Supplier Ted Pella Inc Supplier No. AD11T00X4
Gel-Pak Clear Hinged Box, GEL Retention X4, 2.54cm x 2.54cm x 0.64cm (1 in. x 1 in. x 1/4 in.), 50/PK, Applications: Semiconductors, thin Film Heads, Microwave Devices, Micromachine Sensors, Optoelectronics, Medical Devices