Learn More
Med Vet International Nordic Ice Refrigerant Gel Pack, 5½ x 4 x ¾in, NI16, 36/Case
Catalog No. 503044046
Encompass_Preferred
Refrigerant Gel Pack Nordic Ice For Safe Transport of Food, Pharmaceuticals and Medical Products Ship products ranging from medications and vaccines to exotic meats and chocolate to fresh and frozen foods Nordic Ice gels provide a precise climate ensuring delicate cargo is kept at a proper cold temperature while in transit 5½ x 4 x ¾ inches
Catalog No. 50-304-4046 Supplier Med Vet International Supplier No. 1138686CS
Refrigerant Gel Pack Nordic Ice For Safe Transport of Food, Pharmaceuticals and Medical Products Ship products ranging from medications and vaccines to exotic meats and chocolate to fresh and frozen foods Nordic Ice gels provide a precise climate ensuring delicate cargo is kept at a proper cold temperature while in transit 5½ x 4 x ¾ inches