Learn More
Med Vet International Gel Positioners, Flat Bottom, 16" L, J1282A, Each
Catalog No. 5000581307
Encompass_Preferred
Gel Positioners are designed to aid in positioning patients for either surgery or radiographs. They provide firm but comfortable support. - Made of viscoelastic polymer so it has a soft tissue-like feel - Provides constant support and will not bottom out - Free from latex, silicon, and plasticizer - Will not leak or flow - Radiolucent - Reusable - Easy to clean with standard disinfectants - Compatible with warming blankets - 2 year warranty Flat Bottom Positioner. (Shaped like a loaf of bread) Flat Bottom. 16"L x 4"W x 3 3/4"H Note: Gel positioners are not warranted against damage caused by chewing or nail tears.
Catalog No. 50-005-81307 Supplier Med Vet International Supplier No. JOR1282A
Gel Positioners are designed to aid in positioning patients for either surgery or radiographs. They provide firm but comfortable support. - Made of viscoelastic polymer so it has a soft tissue-like feel - Provides constant support and will not bottom out - Free from latex, silicon, and plasticizer - Will not leak or flow - Radiolucent - Reusable - Easy to clean with standard disinfectants - Compatible with warming blankets - 2 year warranty Flat Bottom Positioner. (Shaped like a loaf of bread) Flat Bottom. 16"L x 4"W x 3 3/4"H Note: Gel positioners are not warranted against damage caused by chewing or nail tears.