Learn More
Plastilite Corp Hot and Cold Packs. Refrigerant Ice Pack. Gel Ice Pack. ControliQ gel pack from Integritemp/Plastilite. Freeze and use as wet ice replacement in chilled and frozen shipments. #TF8, 8oz, 72/cs 5"x5.5"x1.25" nominal size.
Catalog No. NC3579787
Encompass
Hot and Cold Packs. Refrigerant Ice Pack. Gel Ice Pack. ControliQ gel pack from Integritemp/Plastilite. Freeze and use as wet ice replacement in chilled and frozen shipments. Reusable and non-toxic. Certified biodegradable gel material. Plastic film is #4 recyclable LDPE. Packaged in heavy-duty vented boxes for secure shipping & storage and accelerated freezing. #TF8, 8oz, 72/cs 5"x5.5"x1.25 nominal size.
Catalog No. NC3579787 Supplier Plastilite Corp Supplier No. TF8
Hot and Cold Packs. Refrigerant Ice Pack. Gel Ice Pack. ControliQ gel pack from Integritemp/Plastilite. Freeze and use as wet ice replacement in chilled and frozen shipments. Reusable and non-toxic. Certified biodegradable gel material. Plastic film is #4 recyclable LDPE. Packaged in heavy-duty vented boxes for secure shipping & storage and accelerated freezing. #TF8, 8oz, 72/cs 5"x5.5"x1.25 nominal size.