Learn More
Plastilite Corp Hot and Cold Packs. Refrigerant Ice Pack. Gel Ice Pack. ControliQ gel pack from Integritemp/Plastilite. Freeze and use as wet ice replacement in chilled and frozen shipments. #TF16, 16oz, 36/cs 7"x6"x1.25" nominal size.
Catalog No. NC3579790
Encompass
Hot and Cold Packs. Refrigerant Ice Pack. Gel Ice Pack. ControliQ gel pack from Integritemp/Plastilite. Freeze and use as wet ice replacement in chilled and frozen shipments. Reusable and non-toxic. Certified biodegradable gel material. Plastic film is #4 recyclable LDPE. Packaged in heavy-duty vented boxes for secure shipping & storage and accelerated freezing. #TF16, 16oz, 36/cs 7"x6"x1.25" nominal size.
Catalog No. NC3579790 Supplier Plastilite Corp Supplier No. TF16
Hot and Cold Packs. Refrigerant Ice Pack. Gel Ice Pack. ControliQ gel pack from Integritemp/Plastilite. Freeze and use as wet ice replacement in chilled and frozen shipments. Reusable and non-toxic. Certified biodegradable gel material. Plastic film is #4 recyclable LDPE. Packaged in heavy-duty vented boxes for secure shipping & storage and accelerated freezing. #TF16, 16oz, 36/cs 7"x6"x1.25" nominal size.