Learn More
Bio Rad Laboratories Gel Releasers, 5/pk
Catalog No. NC1418739
Encompass
Pkg of 5, gel releaser for Mini-PROTEAN Tetra system, Mini-PROTEAN 3 Dodeca cell, Mini-PROTEAN 3 multi-casting chamber, and PROTEAN Plus Dodeca cell.
This item is not returnable. View return policy
Catalog No. NC1418739 Supplier Bio Rad Laboratories Supplier No. 1653320
Pkg of 5, gel releaser for Mini-PROTEAN Tetra system, Mini-PROTEAN 3 Dodeca cell, Mini-PROTEAN 3 multi-casting chamber, and PROTEAN Plus Dodeca cell.