Learn More
LabScientific Gelatin Coated Self Adhesive Microscope Slides 1" x 3". 1mm thick. Pre-cleaned. Clear with white, frosted edge. Prepared with 2% gelation for use with frozen sections and immunochemistry. 72/pk.
Catalog No. NC9077440
Encompass
Gelatin Coated Self Adhesive Microscope Slides, RNase Free, Positively charged, Clear with white frosted ends, Clean room prepared. 1" x 3". 1mm thick. Pre-cleaned. Clear with white, frosted edge. Prepared with 2% gelation for use with frozen sections and immunochemistry. 72/pk.
Catalog No. NC9077440 Supplier LabScientific Supplier No. 7802
Gelatin Coated Self Adhesive Microscope Slides, RNase Free, Positively charged, Clear with white frosted ends, Clean room prepared. 1" x 3". 1mm thick. Pre-cleaned. Clear with white, frosted edge. Prepared with 2% gelation for use with frozen sections and immunochemistry. 72/pk.