Learn More
Med Vet International Ultrasound Gel Aquasonic Transmission 20 Gram Foil Pouch, 01-01, Each
Catalog No. 502830587
Encompass_Preferred
Overwrapped, sterilized foil pouches Acoustically correct for the broad range of frequencies used in medical ultrasound Completely aqueous, will not stain clothing or damage equipment
Catalog No. 50-283-0587 Supplier Med Vet International Supplier No. 236820EA
Overwrapped, sterilized foil pouches Acoustically correct for the broad range of frequencies used in medical ultrasound Completely aqueous, will not stain clothing or damage equipment