Learn More
Med Vet International Ultrasound Gel Aquasonic Transmission 8.5oz Bottle, 01-08, 12/Box
Supplier:  Med Vet International 1220624BX
Encompass_Preferred
8oz. bottle of Aquasonic Ultrasound Transmission Gel Acoustically correct for the broad range of frequencies used Completely aqueous, will not stain clothing or damage transducers Unique formula is bacteriostatic, non-sensitizing and non-irritating No formaldehyde Not a spermicide Paraben free
Catalog No. 50-282-9967